Learn More
Medchemexpress LLC Tyrphostin 8 | 3785-90-8 | 98.0% | 170.17 g/mol | C10H6N2O | 5 MG
SDP
Catalog No. 5000281925
Encompass_Preferred
Tyrphostin 8 is a small-molecule tyrosine kinase inhibitor supplied for research use. It inhibits EGFR kinase (IC50 = 560 μM), acts as a GTPase inhibitor, and inhibits calcineurin (IC50 = 21 μM). The compound is provided as a high-purity solid and is available in small-mass vials or as a DMSO solution for in vitro biochemical and cell-based assays.
  • High purity (>98%), suitable for in vitro studies.
  • Known activity against EGFR, GTPase, and calcineurin targets.
  • Molecular weight 170.17 g/mol and formula C10H6N2O.
  • Available in multiple small pack sizes and as a 10 mM DMSO solution.
  • Light yellow to yellow solid for handling and storage.
Catalog No. 50-002-81925 Supplier Medchemexpress LLC Supplier No. HYW1742795MG
Tyrphostin 8 is a small-molecule tyrosine kinase inhibitor supplied for research use. It inhibits EGFR kinase (IC50 = 560 μM), acts as a GTPase inhibitor, and inhibits calcineurin (IC50 = 21 μM). The compound is provided as a high-purity solid and is available in small-mass vials or as a DMSO solution for in vitro biochemical and cell-based assays.
  • High purity (>98%), suitable for in vitro studies.
  • Known activity against EGFR, GTPase, and calcineurin targets.
  • Molecular weight 170.17 g/mol and formula C10H6N2O.
  • Available in multiple small pack sizes and as a 10 mM DMSO solution.
  • Light yellow to yellow solid for handling and storage.