Learn More
Medchemexpress LLC Tyrphostin 8 | 3785-90-8 | MFCD00020189 | 98.0% | 170.17 g·mol⁻¹ | C10H6N2O | 1 ML
SDP
Catalog No. 5000283174
Encompass_Preferred
Tyrphostin 8 is a small-molecule inhibitor that targets tyrosine kinases and GTPases and also inhibits calcineurin phosphatase activity. It is supplied at high purity for use in biochemical and cellular assays; chemical identifiers include CAS 3785-90-8, formula C10H6N2O, and molecular weight 170.17 g·mol⁻¹.
Catalog No. 50-002-83174 Supplier Medchemexpress LLC Supplier No. HYW17427910MM/1ML
Tyrphostin 8 is a small-molecule inhibitor that targets tyrosine kinases and GTPases and also inhibits calcineurin phosphatase activity. It is supplied at high purity for use in biochemical and cellular assays; chemical identifiers include CAS 3785-90-8, formula C10H6N2O, and molecular weight 170.17 g·mol⁻¹.