Learn More
Medchemexpress LLC Tyrphostin 8 | 3785-90-8 | MFCD00020189 | 98.0% | 170.17 g/mol | C10H6N2O | 10 MG
SDP
Catalog No. 5000281923
Encompass_Preferred
Tyrphostin 8 is a small-molecule tyrosine kinase inhibitor used as a research reagent to investigate EGFR signaling and related pathways. The compound is supplied as a light yellow to yellow solid with high reported purity and is offered in multiple solid sizes and DMSO solution formats.
  • Inhibits EGFR kinase; reported IC50 ≈ 560 μM
  • Also exhibits GTPase inhibitory activity
  • High purity for reproducible experimental results
  • Available as solid and as 10 mM DMSO solution in multiple sizes
  • Storage recommended: solid at 4°C under nitrogen; solutions at -80°C or -20°C
Catalog No. 50-002-81923 Supplier Medchemexpress LLC Supplier No. HYW17427910MG
Tyrphostin 8 is a small-molecule tyrosine kinase inhibitor used as a research reagent to investigate EGFR signaling and related pathways. The compound is supplied as a light yellow to yellow solid with high reported purity and is offered in multiple solid sizes and DMSO solution formats.
  • Inhibits EGFR kinase; reported IC50 ≈ 560 μM
  • Also exhibits GTPase inhibitory activity
  • High purity for reproducible experimental results
  • Available as solid and as 10 mM DMSO solution in multiple sizes
  • Storage recommended: solid at 4°C under nitrogen; solutions at -80°C or -20°C