Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tyrphostin 8 | 3785-90-8 | MFCD00020189 | 98.0% | 170.17 g/mol | C10H6N2O | 50 MG
SDP

Catalog No. 5000281552
Encompass_Preferred

Tyrphostin 8 is a small-molecule research compound that acts as a weak epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor and a more potent inhibitor of the serine/threonine phosphatase calcineurin. Supplied as a solid for laboratory research, it is intended for research use only.

  • Small-molecule inhibitor of EGFR and calcineurin (EGFR IC50 560 μM; calcineurin IC50 21 μM).
  • High purity: 98.0%.
  • Chemical formula C10H6N2O; molecular weight 170.17 g/mol.
  • CAS number 3785-90-8.
  • Soluble in DMSO at ≥ 100 mg/mL.
  • Appearance: light yellow to yellow solid.
  • Storage: store at 4°C under nitrogen; in solvent store at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical applications.

Catalog No. 50-002-81552 Supplier Medchemexpress LLC Supplier No. HYW17427950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tyrphostin 8 is a small-molecule research compound that acts as a weak epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor and a more potent inhibitor of the serine/threonine phosphatase calcineurin. Supplied as a solid for laboratory research, it is intended for research use only.

  • Small-molecule inhibitor of EGFR and calcineurin (EGFR IC50 560 μM; calcineurin IC50 21 μM).
  • High purity: 98.0%.
  • Chemical formula C10H6N2O; molecular weight 170.17 g/mol.
  • CAS number 3785-90-8.
  • Soluble in DMSO at ≥ 100 mg/mL.
  • Appearance: light yellow to yellow solid.
  • Storage: store at 4°C under nitrogen; in solvent store at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.