Learn More
Apexbio Technology LLC Tyrphostin 9 10537-47-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063768
Encompass_Preferred
Tyrphostin 9 is a selective inhibitor targeting platelet-derived growth factor receptor (PDGFR) and epidermal growth factor receptor (EGFR) It inhibits PDGFR autophosphorylation with an IC50 of approximately 2 5 M and EGFR kinase activity (IC50 around 460 M) Additionally Tyrphostin 9 inhibits platelet-derived growth factor (PDGF)-stimulated smooth muscle cell proliferation (IC50 approximately 40 nM) as well as phosphorylation of phospholipase C gamma (PLC IC50 2 5 M) and induction of PDGF-dependent c-fos expression In research models Tyrphostin 9 has been utilized to study receptor-mediated signal transduction and vascular injury-related neointimal proliferation processes
Catalog No. 50-000-63768 Supplier Apexbio Technology LLC Supplier No. B14945.1
Tyrphostin 9 is a selective inhibitor targeting platelet-derived growth factor receptor (PDGFR) and epidermal growth factor receptor (EGFR) It inhibits PDGFR autophosphorylation with an IC50 of approximately 2 5 M and EGFR kinase activity (IC50 around 460 M) Additionally Tyrphostin 9 inhibits platelet-derived growth factor (PDGF)-stimulated smooth muscle cell proliferation (IC50 approximately 40 nM) as well as phosphorylation of phospholipase C gamma (PLC IC50 2 5 M) and induction of PDGF-dependent c-fos expression In research models Tyrphostin 9 has been utilized to study receptor-mediated signal transduction and vascular injury-related neointimal proliferation processes