Learn More
Selleck Chemical LLC Tyrphostin 9 S2895-25mg
SDP
Catalog No. 503111843
Encompass_Preferred
Tyrphostin 9 (SF 6847 RG-50872) is firstly designed as an EGFR inhibitor with IC50 of 460 M but is also found to be more potent to PDGFR with IC50 of 0 5 M
Catalog No. 50-311-1843 Supplier Selleck Chemical LLC Supplier No. S289525MG
Tyrphostin 9 (SF 6847 RG-50872) is firstly designed as an EGFR inhibitor with IC50 of 460 M but is also found to be more potent to PDGFR with IC50 of 0 5 M