Learn More
Enzo Life Sciences Tyrphostin 9 (50mg). CAS: 10537-47-0
SDP
Supplier:  Enzo Life Sciences BMLEI2150050
Encompass_Preferred
Tyrphostin 9 is a selective inhibitor of the PDGF receptor tyrosine kinase (IC50=1.2 µM). It is also a potent (10 nM) uncoupler of oxidative phosphorylation. Alternative name: RG-50872, Malonaben, SF 6847. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO (>25mg/ml) or ethanol (20mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0612