Learn More
Medchemexpress LLC Tyrphostin A1 | 2826-26-8 | 99.0% | 184.19 | 100 MG
SDP
Catalog No. 5000394253
Encompass_Preferred
Tyrphostin A1 inhibits CD40L-stimulated IL-12 production in macrophage cultures and antigen-induced generation of Th1 cells. It causes a dose-dependent decrease of IL-12 p40 and blocks CD40L-induced translocation of NF-κB to the nucleus, reducing activation of the IL-12 p40 gene. This compound is a weaker inhibitor of TK, often used to differentiate TK-mediated effects from other non-specific effects.
  • Inhibits CD40L-stimulated IL-12 production
  • Reduces antigen-induced generation of Th1 cells
  • Blocks CD40L-induced translocation of NF-κB to the nucleus
  • Attenuates experimental allergic encephalomyelitis (EAE) in SJL/J mice
  • Useful for differentiating TK-mediated effects from non-specific effects
  • For research use only
Catalog No. 50-003-94253 Supplier Medchemexpress LLC Supplier No. HY16668100MG
Tyrphostin A1 inhibits CD40L-stimulated IL-12 production in macrophage cultures and antigen-induced generation of Th1 cells. It causes a dose-dependent decrease of IL-12 p40 and blocks CD40L-induced translocation of NF-κB to the nucleus, reducing activation of the IL-12 p40 gene. This compound is a weaker inhibitor of TK, often used to differentiate TK-mediated effects from other non-specific effects.
  • Inhibits CD40L-stimulated IL-12 production
  • Reduces antigen-induced generation of Th1 cells
  • Blocks CD40L-induced translocation of NF-κB to the nucleus
  • Attenuates experimental allergic encephalomyelitis (EAE) in SJL/J mice
  • Useful for differentiating TK-mediated effects from non-specific effects
  • For research use only