Learn More
Medchemexpress LLC Tyrphostin A1 | 2826-26-8 | 99.0% | 184.19 | 25 MG
SDP
Catalog No. 5000395182
Encompass_Preferred
Tyrphostin A1 (AG9) is a compound that inhibits CD40L-stimulated IL-12 production and antigen-induced generation of Th1 cells. It blocks CD40L-induced translocation of NF-κB to the nucleus, reducing IL-12 p40 gene activation. In vivo, it decreases the generation of myelin basic protein (MBP) specific encephalitogenic T cells and attenuates experimental allergic encephalomyelitis (EAE).
  • Inhibits CD40L-stimulated IL-12 production in macrophage cultures
  • Inhibits antigen-induced generation of Th1 cells
  • Blocks CD40L-induced translocation of NF-κB to the nucleus
  • Reduces activation of the IL-12 p40 gene
  • Decreases generation of myelin basic protein (MBP) specific encephalitogenic T cells
  • Attenuates experimental allergic encephalomyelitis (EAE)
  • Differentiates TK-mediated effects from non-specific effects due to weak TK inhibition (IC50>1250 μM for epidermal growth factor receptor (EGFR) kinase)
Catalog No. 50-003-95182 Supplier Medchemexpress LLC Supplier No. 25MGHY16668
Tyrphostin A1 (AG9) is a compound that inhibits CD40L-stimulated IL-12 production and antigen-induced generation of Th1 cells. It blocks CD40L-induced translocation of NF-κB to the nucleus, reducing IL-12 p40 gene activation. In vivo, it decreases the generation of myelin basic protein (MBP) specific encephalitogenic T cells and attenuates experimental allergic encephalomyelitis (EAE).
  • Inhibits CD40L-stimulated IL-12 production in macrophage cultures
  • Inhibits antigen-induced generation of Th1 cells
  • Blocks CD40L-induced translocation of NF-κB to the nucleus
  • Reduces activation of the IL-12 p40 gene
  • Decreases generation of myelin basic protein (MBP) specific encephalitogenic T cells
  • Attenuates experimental allergic encephalomyelitis (EAE)
  • Differentiates TK-mediated effects from non-specific effects due to weak TK inhibition (IC50>1250 μM for epidermal growth factor receptor (EGFR) kinase)