Learn More
Medchemexpress LLC HY-15511 100mg , Tyrphostin A9 CAS:10537-47-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15511100MG
Encompass_Preferred
HY-15511 100mg Tyrphostin A9 CAS: 10537-47-0 Tyrphostin A9, a PDGFR inhibitor, is a potent inducer of mitochondrial fission. Tyrphostin A9 emerged as the most potent and selective of 51 tyrosine kinase inhibitors tested against the TNF-induced respiratory burst. Tyrphostin A9 has anti-influenza virus activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9751