Learn More
Medchemexpress LLC Tyrphostin A9 | 10537-47-0 | MFCD00209853 | 99.9% | 282.38 g/mol | C18H22N2O | 10 MG
SDP
Catalog No. 5000287357
Encompass_Preferred
Tyrphostin A9 is a small-molecule PDGFR inhibitor provided as an analytical standard for research applications. It is used in biochemical and cell-based studies to investigate tyrosine kinase signaling and mitochondrial dynamics. The material is supplied with purity and storage information and is available in multiple solid and solution formats for laboratory use.
  • High purity suitable for analytical and research use.
  • Useful as a PDGFR and tyrosine kinase inhibitor in cell and biochemical assays.
  • Available in solid and solution formats for flexible experimental workflows.
  • Documented stability when stored under recommended conditions.
  • Supplied with supporting documentation such as a certificate of analysis or data sheet.
Catalog No. 50-002-87357 Supplier Medchemexpress LLC Supplier No. HY1551110MG
Tyrphostin A9 is a small-molecule PDGFR inhibitor provided as an analytical standard for research applications. It is used in biochemical and cell-based studies to investigate tyrosine kinase signaling and mitochondrial dynamics. The material is supplied with purity and storage information and is available in multiple solid and solution formats for laboratory use.
  • High purity suitable for analytical and research use.
  • Useful as a PDGFR and tyrosine kinase inhibitor in cell and biochemical assays.
  • Available in solid and solution formats for flexible experimental workflows.
  • Documented stability when stored under recommended conditions.
  • Supplied with supporting documentation such as a certificate of analysis or data sheet.