Learn More
Enzo Life Sciences Tyrphostin AG 1024 (5mg). CAS: 65678-07-1
SDP
Catalog No. 502008435
Encompass_Preferred
Specific inhibitor of insulin-like growth factor-1 (IGF-1) and insulin receptor kinases. Induces apoptosis by downregulation of the expression of phospho-Akt1 and Bcl-2. Also increases Bax, p53 and p21 expression. Formula: C14H13BrN2O. MW: 305.2. Purity: ≥98% (HPLC (UV)). Solubility: Soluble in acetonitrile (5mg/ml), DMSO (8mg/ml), dichloromethane (23mg/ml) or methanol (5mg/ml); insoluble in 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-8435 Supplier Enzo Life Sciences Supplier No. ALX270217M005
Specific inhibitor of insulin-like growth factor-1 (IGF-1) and insulin receptor kinases. Induces apoptosis by downregulation of the expression of phospho-Akt1 and Bcl-2. Also increases Bax, p53 and p21 expression. Formula: C14H13BrN2O. MW: 305.2. Purity: ≥98% (HPLC (UV)). Solubility: Soluble in acetonitrile (5mg/ml), DMSO (8mg/ml), dichloromethane (23mg/ml) or methanol (5mg/ml); insoluble in 100% ethanol. Long Term Storage: -20°C.