Learn More
Medchemexpress LLC Tyrphostin AG 112 | 144978-82-5 | 98.3% | 236.23 | 10 MG
SDP
Catalog No. 5000437105
Encompass_Preferred
Tyrphostin AG 112 is an EGFR phosphorylation inhibitor, intended for research use only.
  • Inhibits EGFR phosphorylation.
  • Detailed solubility information for in vitro and in vivo applications is available.
  • Comprehensive documentation including Data Sheet, COA, SDS, HNMR, LCMS, and Handling Instructions.
  • Target: EGFR.
  • Pathway: JAK/STAT Signaling; Protein Tyrosine Kinase/RTK.
Catalog No. 50-004-37105 Supplier Medchemexpress LLC Supplier No. HY11247410MG
Tyrphostin AG 112 is an EGFR phosphorylation inhibitor, intended for research use only.
  • Inhibits EGFR phosphorylation.
  • Detailed solubility information for in vitro and in vivo applications is available.
  • Comprehensive documentation including Data Sheet, COA, SDS, HNMR, LCMS, and Handling Instructions.
  • Target: EGFR.
  • Pathway: JAK/STAT Signaling; Protein Tyrosine Kinase/RTK.