Learn More
Enzo Life Sciences Tyrphostin AG 1295 (5mg). CAS: 71897-07-9
SDP
Catalog No. 502008375
Encompass_Preferred
PDGF inhibitor. Selective inhibitor of tyrosine kinase in platelet-derived growth factor (PDGF) receptor. Alternative name: 6,7-Dimethyl-2-phenylquinoxaline. Formula: C16H14N2. MW: 234.3. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C. Handling: Store tightly sealed in the dark. Protect from light.
Catalog No. 50-200-8375 Supplier Enzo Life Sciences Supplier No. ALX270035M005
PDGF inhibitor. Selective inhibitor of tyrosine kinase in platelet-derived growth factor (PDGF) receptor. Alternative name: 6,7-Dimethyl-2-phenylquinoxaline. Formula: C16H14N2. MW: 234.3. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C. Handling: Store tightly sealed in the dark. Protect from light.