Learn More
Apexbio Technology LLC Tyrphostin AG 1296 146535-11-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062999
Encompass_Preferred
Tyrphostin AG 1296 (CAS 146535-11-7) is a selective inhibitor of platelet-derived growth factor receptor (PDGFR) kinase via ATP-competitive binding Its inhibitory activity (IC50 0 3 0 5 M) results from inducing conformational changes in the ATP-binding domain of PDGFR blocking receptor autophosphorylation induced by ligand binding In vitro studies demonstrate inhibition of PDGF-stimulated mitogenesis (IC50 1 5 M) and cell proliferation (IC50 3 2 M) using Swiss 3T3 cells without significant effects on EGF receptor signaling at concentrations up to 100 M Tyrphostin AG 1296 serves as a research tool for investigating PDGFR-mediated signaling pathways
Catalog No. 50-000-62999 Supplier Apexbio Technology LLC Supplier No. A24775.1
Tyrphostin AG 1296 (CAS 146535-11-7) is a selective inhibitor of platelet-derived growth factor receptor (PDGFR) kinase via ATP-competitive binding Its inhibitory activity (IC50 0 3 0 5 M) results from inducing conformational changes in the ATP-binding domain of PDGFR blocking receptor autophosphorylation induced by ligand binding In vitro studies demonstrate inhibition of PDGF-stimulated mitogenesis (IC50 1 5 M) and cell proliferation (IC50 3 2 M) using Swiss 3T3 cells without significant effects on EGF receptor signaling at concentrations up to 100 M Tyrphostin AG 1296 serves as a research tool for investigating PDGFR-mediated signaling pathways