Learn More
Selleck Chemical LLC Tyrphostin AG 1296
SDP
Catalog No. 5000000209
Encompass_Preferred
Tyrphostin AG 1296 is an inhibitor of PDGFR with IC50 of 0 3-0 5 M no activity to EGFR Tyrphostin AG1296 inhibits FGFR and c-Kit with IC50 of 12 3 M and 1 8 M in Swiss 3T3 cells Tyrphostin AG1296 induces dramatic apoptosis in A375R cells
Catalog No. 50-000-00209 Supplier Selleck Chemical LLC Supplier No. S80245MG
Tyrphostin AG 1296 is an inhibitor of PDGFR with IC50 of 0 3-0 5 M no activity to EGFR Tyrphostin AG1296 inhibits FGFR and c-Kit with IC50 of 12 3 M and 1 8 M in Swiss 3T3 cells Tyrphostin AG1296 induces dramatic apoptosis in A375R cells