Learn More
Enzo Life Sciences Tyrphostin AG 527 (25mg). CAS: 133550-32-0
SDP
Catalog No. 502008413
Encompass_Preferred
Alternative name: Tyrphostin B44, (-)-(R)-N-(alpha-Methylbenzyl)-3,4-dihydroxybenzylidenecyanoacetamide. Formula: C18H16N2O3. MW: 308.3. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO or 100% ethanol. Long Term Storage: -20°C. Handling: Store tightly sealed in the dark. Protect from light.
Catalog No. 50-200-8413 Supplier Enzo Life Sciences Supplier No. ALX270141M025
Alternative name: Tyrphostin B44, (-)-(R)-N-(alpha-Methylbenzyl)-3,4-dihydroxybenzylidenecyanoacetamide. Formula: C18H16N2O3. MW: 308.3. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO or 100% ethanol. Long Term Storage: -20°C. Handling: Store tightly sealed in the dark. Protect from light.