Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tyrphostin AG 528 | 133550-49-9 | MFCD01076474 | 98.0% | 306.32 | C18H14N2O3 | 10 MG
SDP

Catalog No. 5000214069
Encompass_Preferred

Tyrphostin AG 528 is a small-molecule protein tyrosine kinase inhibitor used as a research reagent for biochemical and cellular studies targeting EGFR and ErbB2. It is supplied as a solid with high purity and defined storage recommendations for long-term stability.

  • Small-molecule inhibitor selective for EGFR and ErbB2.
  • Reported IC50: EGFR 4.9 μM, ErbB2 2.1 μM.
  • High purity (98.0%) suitable for preclinical research.
  • Solid, light yellow to yellow appearance for easy handling.
  • Molecular weight 306.32 and formula C18H14N2O3 for characterization.
  • Available in small pack sizes for screening and assay development.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years).

Catalog No. 50-002-14069 Supplier Medchemexpress LLC Supplier No. HY10049910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tyrphostin AG 528 is a small-molecule protein tyrosine kinase inhibitor used as a research reagent for biochemical and cellular studies targeting EGFR and ErbB2. It is supplied as a solid with high purity and defined storage recommendations for long-term stability.

  • Small-molecule inhibitor selective for EGFR and ErbB2.
  • Reported IC50: EGFR 4.9 μM, ErbB2 2.1 μM.
  • High purity (98.0%) suitable for preclinical research.
  • Solid, light yellow to yellow appearance for easy handling.
  • Molecular weight 306.32 and formula C18H14N2O3 for characterization.
  • Available in small pack sizes for screening and assay development.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.