Learn More
Medchemexpress LLC Tyrphostin AG 528 | 133550-49-9 | MFCD01076474 | 98.0% | 306.32 g·mol^-1 | C18H14N2O3 | 25 MG
SDP
Catalog No. 5000214736
Encompass_Preferred
Tyrphostin AG 528 is a small-molecule protein tyrosine kinase inhibitor used as a research reagent to probe EGFR and ErbB2 (HER2) signaling pathways. It is supplied as a high-purity solid for biochemical and cellular studies.
  • Inhibits EGFR and ErbB2 with reported IC50s of 4.9 μM and 2.1 μM.
  • High purity (98.0%) suitable for laboratory research.
  • Supplied as a solid, light yellow to yellow in color.
  • Molecular weight 306.32 g·mol⁻¹; formula C18H14N2O3.
  • Available in small research quantities, including 25 mg packaging.
Catalog No. 50-002-14736 Supplier Medchemexpress LLC Supplier No. HY10049925MG
Tyrphostin AG 528 is a small-molecule protein tyrosine kinase inhibitor used as a research reagent to probe EGFR and ErbB2 (HER2) signaling pathways. It is supplied as a high-purity solid for biochemical and cellular studies.
  • Inhibits EGFR and ErbB2 with reported IC50s of 4.9 μM and 2.1 μM.
  • High purity (98.0%) suitable for laboratory research.
  • Supplied as a solid, light yellow to yellow in color.
  • Molecular weight 306.32 g·mol⁻¹; formula C18H14N2O3.
  • Available in small research quantities, including 25 mg packaging.