Learn More
Medchemexpress LLC Tyrphostin AG 528 | 133550-49-9 | MFCD01076474 | 98.0% | 306.32 g/mol | C18H14N2O3 | 5 MG
SDP
Catalog No. 5000214741
Encompass_Preferred
Tyrphostin AG 528 is a small-molecule tyrosine kinase inhibitor of EGFR and ErbB2 used in preclinical research to probe receptor signaling. It is supplied as a purified research chemical for in vitro and exploratory studies; chemical formula C18H14N2O3, molecular weight 306.32 g/mol, purity 98.0%.
  • Potent inhibitor of EGFR and ErbB2 with reported IC50s of 4.9 μM and 2.1 μM.
  • High purity suitable for biochemical and cellular assays.
  • Available in small quantities appropriate for dose-response studies.
  • Contains the CAS number 133550-49-9 for unambiguous identification.
  • Supplied with a manufacturer datasheet detailing physicochemical properties and handling.
Catalog No. 50-002-14741 Supplier Medchemexpress LLC Supplier No. HY1004995MG
Tyrphostin AG 528 is a small-molecule tyrosine kinase inhibitor of EGFR and ErbB2 used in preclinical research to probe receptor signaling. It is supplied as a purified research chemical for in vitro and exploratory studies; chemical formula C18H14N2O3, molecular weight 306.32 g/mol, purity 98.0%.
  • Potent inhibitor of EGFR and ErbB2 with reported IC50s of 4.9 μM and 2.1 μM.
  • High purity suitable for biochemical and cellular assays.
  • Available in small quantities appropriate for dose-response studies.
  • Contains the CAS number 133550-49-9 for unambiguous identification.
  • Supplied with a manufacturer datasheet detailing physicochemical properties and handling.