Learn More
Selleck Chemical LLC Tyrphostin AG-528
SDP
Catalog No. 5000002327
Encompass_Preferred
Tyrphostin AG-528 (Tyrphostin B66) is a potent inhibitor of epidermal growth factor receptors (EGFR) and ErbB2/HER2 with IC50 of 4 9 M and 2 1 M respectively Tyrphostin AG-528 exhibits anticancer activity
Catalog No. 50-000-02327 Supplier Selleck Chemical LLC Supplier No. S68055MG
Tyrphostin AG-528 (Tyrphostin B66) is a potent inhibitor of epidermal growth factor receptors (EGFR) and ErbB2/HER2 with IC50 of 4 9 M and 2 1 M respectively Tyrphostin AG-528 exhibits anticancer activity