Learn More
TARGETMOL CHEMICALS INC Tyrphostin AG 528 25MG
SDP
Catalog No. 502878685
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Tyrphostin AG 528 (Tyrphostin B66) is an inhibitor of EGFR (IC50 4.9 uM) and ErbB2 (IC50 2.1 uM). Purity 99.74%
Catalog No. 50-287-8685 Supplier TARGETMOL CHEMICALS INC Supplier No. T452825MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Tyrphostin AG 528 (Tyrphostin B66) is an inhibitor of EGFR (IC50 4.9 uM) and ErbB2 (IC50 2.1 uM). Purity 99.74%