Learn More
TARGETMOL CHEMICALS INC TYRPHOSTIN AG 538 5MG
SDP
Catalog No. 502579278
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Tyrphostin AG 538 is a chalcone compound an IGF-1 receptor kinase inhibitor (IC₅0 for 400 nM) that is potent cell-permeable reversible and competitive. purity: 99%
Catalog No. 50-257-9278 Supplier TARGETMOL CHEMICALS INC Supplier No. T677075MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Tyrphostin AG 538 is a chalcone compound an IGF-1 receptor kinase inhibitor (IC₅0 for 400 nM) that is potent cell-permeable reversible and competitive. purity: 99%