Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tyrphostin AG-825 | 149092-50-2 | MFCD01074971 | 98.0% | 397.47 g·mol⁻¹ | C19H15N3O3S2 | 10 MG
SDP

Catalog No. 5000288386
Encompass_Preferred

AG-825 (Tyrphostin AG-825) is a selective, ATP-competitive ErbB2 (HER2) tyrosine kinase inhibitor used in cellular and biochemical research to probe ErbB2 signaling and cell proliferation. It exhibits anticancer and anti-inflammatory activity and has been shown to accelerate apoptosis in human neutrophils.

  • Selective ErbB2 inhibitor with reported IC50 of 0.35 μM.
  • Additional activity against EGFR with IC50 ~19 μM.
  • High purity suitable for research, reported at 98.02%.
  • Molecular weight 397.47 g·mol⁻¹; formula C19H15N3O3S2.
  • Soluble in DMSO (recommended 250 mg/mL with ultrasonic assistance) and formulatable for in vivo use.
  • Available in small research pack sizes and larger quantities for expanded studies.
  • Stable as powder and in solvent when stored under the recommended conditions.

Catalog No. 50-002-88386 Supplier Medchemexpress LLC Supplier No. HY1584410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AG-825 (Tyrphostin AG-825) is a selective, ATP-competitive ErbB2 (HER2) tyrosine kinase inhibitor used in cellular and biochemical research to probe ErbB2 signaling and cell proliferation. It exhibits anticancer and anti-inflammatory activity and has been shown to accelerate apoptosis in human neutrophils.

  • Selective ErbB2 inhibitor with reported IC50 of 0.35 μM.
  • Additional activity against EGFR with IC50 ~19 μM.
  • High purity suitable for research, reported at 98.02%.
  • Molecular weight 397.47 g·mol⁻¹; formula C19H15N3O3S2.
  • Soluble in DMSO (recommended 250 mg/mL with ultrasonic assistance) and formulatable for in vivo use.
  • Available in small research pack sizes and larger quantities for expanded studies.
  • Stable as powder and in solvent when stored under the recommended conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.