Learn More
Apexbio Technology LLC Tyrphostin AG 879, 10mg. Cas: 148741-30-4 MFCD: MFCD00236450. HER2 inhibitor
SDP
Supplier:  Apexbio Technology LLC B154410
Encompass_Preferred
Tyrphostin AG879 is a tyrosine kinase inhibitor that inhibits the phosphorylation of TrKA, but not TrKB and TrKC. Tyrphostin AG879 is also a ErbB2 kinase inhibitor, has at least 500-fold higher selectivity to ErbB2 than EGFR (IC50 > 500 mol/L). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5255