Learn More
Medchemexpress LLC Tyrphostin AG 879 | 148741-30-4 | MFCD00236450 | 99.3% | 10 MG
SDP
Catalog No. 5000297537
Encompass_Preferred
Tyrphostin AG 879 is a small-molecule tyrosine kinase inhibitor widely used in laboratory research. It shows selectivity for ErbB2 (HER2) with a reported IC50 of about 1 μM and reduces TrkA phosphorylation at higher micromolar concentrations. The compound is supplied as a powder, is soluble in DMSO, and is intended for in vitro signaling and kinase pathway studies.
Catalog No. 50-002-97537 Supplier Medchemexpress LLC Supplier No. HY2087810MG
Tyrphostin AG 879 is a small-molecule tyrosine kinase inhibitor widely used in laboratory research. It shows selectivity for ErbB2 (HER2) with a reported IC50 of about 1 μM and reduces TrkA phosphorylation at higher micromolar concentrations. The compound is supplied as a powder, is soluble in DMSO, and is intended for in vitro signaling and kinase pathway studies.