Learn More
Selleck Chemical LLC Tyrphostin AG 879 S2816-5mg
SDP
Catalog No. 503109025
Encompass_Preferred
Tyrphostin AG 879 potently inhibits HER2/ErbB2 with IC50 of 1 M 100- and 500-fold higher selective to ErbB2 than PDGFR and EGFR
Catalog No. 50-310-9025 Supplier Selleck Chemical LLC Supplier No. S28165MG
Tyrphostin AG 879 potently inhibits HER2/ErbB2 with IC50 of 1 M 100- and 500-fold higher selective to ErbB2 than PDGFR and EGFR