Learn More
TARGETMOL CHEMICALS INC TYRPHOSTIN AG1296 10MG
SDP
Catalog No. 502579301
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tyrphostin AG1296 (Tyrphostin AG 1296) is an inhibitor of PDGFR with IC50 of 0.3-0.5 uM no activity to EGFR. purity: 99%
Catalog No. 50-257-9301 Supplier TARGETMOL CHEMICALS INC Supplier No. T671110MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tyrphostin AG1296 (Tyrphostin AG 1296) is an inhibitor of PDGFR with IC50 of 0.3-0.5 uM no activity to EGFR. purity: 99%