Learn More
Medchemexpress LLC Tyrphostin AG1296 | 146535-11-7 | 99.69% | 266.29 | 5 MG
SDP
Catalog No. 5000364654
Encompass_Preferred
Tyrphostin AG1296 is a potent and selective inhibitor of platelet-derived growth factor receptor (PDGFR) with an IC50 of 0.8 μM. It inhibits the signaling of human PDGF α- and β-receptors, the related stem cell factor receptor (c-Kit), and FLT3.
  • Potent and selective inhibitor of PDGFR with an IC50 of 0.8 μM.
  • Inhibits signaling of human PDGF α- and β-receptors.
  • Inhibits the related stem cell factor receptor (c-Kit).
  • Potent inhibitor of FLT3 in the micromolar range.
  • Suppresses viability of PLX4032-resistant melanoma cells.
  • Induces apoptosis of A375R cells.
  • Inhibits PDGFR phosphorylation.
  • Inhibits migration of A375R cells.
  • Suppresses tumor growth in mice models.
  • Inhibits atherosclerotic plaque progression and enhances plaque stability.
Catalog No. 50-003-64654 Supplier Medchemexpress LLC Supplier No. 5MGHY13894
Tyrphostin AG1296 is a potent and selective inhibitor of platelet-derived growth factor receptor (PDGFR) with an IC50 of 0.8 μM. It inhibits the signaling of human PDGF α- and β-receptors, the related stem cell factor receptor (c-Kit), and FLT3.
  • Potent and selective inhibitor of PDGFR with an IC50 of 0.8 μM.
  • Inhibits signaling of human PDGF α- and β-receptors.
  • Inhibits the related stem cell factor receptor (c-Kit).
  • Potent inhibitor of FLT3 in the micromolar range.
  • Suppresses viability of PLX4032-resistant melanoma cells.
  • Induces apoptosis of A375R cells.
  • Inhibits PDGFR phosphorylation.
  • Inhibits migration of A375R cells.
  • Suppresses tumor growth in mice models.
  • Inhibits atherosclerotic plaque progression and enhances plaque stability.