Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tyrphostin AG1433 | 168835-90-3 | 99.1% | 1 ML
SDP

Catalog No. 5000233125
Encompass_Preferred

Tyrphostin AG1433 is a small-molecule tyrosine kinase inhibitor selective for PDGFRβ, supplied as a 10 mM solution in DMSO (1 mL) for research use. The compound has molecular formula C16H14N2O2 and molecular weight 266.29 g·mol⁻¹.

  • Supplied as a 10 mM solution in DMSO, 1 mL.
  • High purity (≈99.1%) suitable for biological research.
  • Molecular formula C16H14N2O2; molecular weight 266.29 g·mol⁻¹.
  • Also available as solid in multiple milligram quantities for custom preparations.
  • For research use only; not for human or veterinary applications.

Catalog No. 50-002-33125 Supplier Medchemexpress LLC Supplier No. HY11975710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tyrphostin AG1433 is a small-molecule tyrosine kinase inhibitor selective for PDGFRβ, supplied as a 10 mM solution in DMSO (1 mL) for research use. The compound has molecular formula C16H14N2O2 and molecular weight 266.29 g·mol⁻¹.

  • Supplied as a 10 mM solution in DMSO, 1 mL.
  • High purity (≈99.1%) suitable for biological research.
  • Molecular formula C16H14N2O2; molecular weight 266.29 g·mol⁻¹.
  • Also available as solid in multiple milligram quantities for custom preparations.
  • For research use only; not for human or veterinary applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.