Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tyrphostin AG1433 | 168835-90-3 | 99.1 % | 100 MG
SDP

Catalog No. 5000233326
Encompass_Preferred

Tyrphostin AG1433 is a small-molecule tyrosine kinase inhibitor selective for PDGFRβ and VEGFR-2 (Flk-1/KDR). Supplied as a solid for research use, it is used to study receptor-mediated signaling and angiogenesis.

  • Selective inhibitor of PDGFRβ and VEGFR-2.
  • Reported IC50 values: PDGFRβ 5.0 μM; VEGFR-2 9.3 μM.
  • Approximately 99.1% purity (HPLC).
  • Available in multiple solid pack sizes, including 100 mg.
  • Stable when stored under recommended conditions for powder (-20°C or 4°C).

Catalog No. 50-002-33326 Supplier Medchemexpress LLC Supplier No. HY119757100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tyrphostin AG1433 is a small-molecule tyrosine kinase inhibitor selective for PDGFRβ and VEGFR-2 (Flk-1/KDR). Supplied as a solid for research use, it is used to study receptor-mediated signaling and angiogenesis.

  • Selective inhibitor of PDGFRβ and VEGFR-2.
  • Reported IC50 values: PDGFRβ 5.0 μM; VEGFR-2 9.3 μM.
  • Approximately 99.1% purity (HPLC).
  • Available in multiple solid pack sizes, including 100 mg.
  • Stable when stored under recommended conditions for powder (-20°C or 4°C).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.