Learn More
Medchemexpress LLC Tyrphostin AG1433 | 168835-90-3 | 99.1% | 5 MG
SDP
Catalog No. 5000234625
Encompass_Preferred
Tyrphostin AG1433 is a small-molecule tyrosine kinase inhibitor used in preclinical research to selectively inhibit PDGFRβ and VEGFR-2. It is supplied as a high-purity research chemical suitable for biochemical and cell-based assays, with documented solubility in DMSO and common in vivo vehicle formulations.
  • High purity suitable for sensitive assays.
  • Molecular weight 266.29 g/mol and formula C16H14N2O2.
  • High DMSO solubility (~62.5 mg/mL) and in vivo vehicle solubility ≥ 2.08 mg/mL.
  • Useful for probing PDGFR and VEGFR signaling in vitro and in vivo models.
  • Typically stored as a powder at low temperature to preserve stability.
Catalog No. 50-002-34625 Supplier Medchemexpress LLC Supplier No. HY1197575MG
Tyrphostin AG1433 is a small-molecule tyrosine kinase inhibitor used in preclinical research to selectively inhibit PDGFRβ and VEGFR-2. It is supplied as a high-purity research chemical suitable for biochemical and cell-based assays, with documented solubility in DMSO and common in vivo vehicle formulations.
  • High purity suitable for sensitive assays.
  • Molecular weight 266.29 g/mol and formula C16H14N2O2.
  • High DMSO solubility (~62.5 mg/mL) and in vivo vehicle solubility ≥ 2.08 mg/mL.
  • Useful for probing PDGFR and VEGFR signaling in vitro and in vivo models.
  • Typically stored as a powder at low temperature to preserve stability.