Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tyrphostin AG1433 | 168835-90-3 | 99.1% | 50 MG
SDP

Catalog No. 5000234260
Encompass_Preferred

Tyrphostin AG1433 is a small-molecule tyrosine kinase inhibitor that selectively targets PDGFRβ and VEGFR-2 and is used as an angiogenesis inhibitor in research. For research use only; not for human or clinical use.

  • Selective PDGFRβ and VEGFR-2 inhibitor
  • Reported IC50 values: PDGFRβ ~5.0 μM; VEGFR-2 ~9.3 μM
  • Molecular formula C16H14N2O2; molecular weight 266.29 g/mol
  • High purity, typically 99.1% by HPLC
  • Available as solid in small pack sizes (for example, 50 mg) and as DMSO solutions
  • Provided with supporting documents: datasheet, SDS, COA, and LCMS

Catalog No. 50-002-34260 Supplier Medchemexpress LLC Supplier No. HY11975750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tyrphostin AG1433 is a small-molecule tyrosine kinase inhibitor that selectively targets PDGFRβ and VEGFR-2 and is used as an angiogenesis inhibitor in research. For research use only; not for human or clinical use.

  • Selective PDGFRβ and VEGFR-2 inhibitor
  • Reported IC50 values: PDGFRβ ~5.0 μM; VEGFR-2 ~9.3 μM
  • Molecular formula C16H14N2O2; molecular weight 266.29 g/mol
  • High purity, typically 99.1% by HPLC
  • Available as solid in small pack sizes (for example, 50 mg) and as DMSO solutions
  • Provided with supporting documents: datasheet, SDS, COA, and LCMS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.