Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tyrphostin AG30 | 122520-79-0 | MFCD09739079 | 99.7% | 205.17 | C10H7NO4 | 1 ML
SDP

Catalog No. 5000228595
Encompass_Preferred

Tyrphostin AG30 (CAS 122520-79-0) is a small-molecule inhibitor of EGFR tyrosine kinase supplied as a 10 mM solution in DMSO (1 mL), intended for biochemical and cell-based research.

  • Potent and selective EGFR tyrosine kinase inhibition.
  • Supplied as a ready-to-use 10 mM solution in DMSO.
  • High purity suitable for research applications.
  • Molecular weight 205.17 and formula C10H7NO4 provided for characterization.
  • Storage recommendations provided to maintain compound stability.

Catalog No. 50-002-28595 Supplier Medchemexpress LLC Supplier No. HY11853210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tyrphostin AG30 (CAS 122520-79-0) is a small-molecule inhibitor of EGFR tyrosine kinase supplied as a 10 mM solution in DMSO (1 mL), intended for biochemical and cell-based research.

  • Potent and selective EGFR tyrosine kinase inhibition.
  • Supplied as a ready-to-use 10 mM solution in DMSO.
  • High purity suitable for research applications.
  • Molecular weight 205.17 and formula C10H7NO4 provided for characterization.
  • Storage recommendations provided to maintain compound stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.