Learn More
Medchemexpress LLC HY-118532 5mg Medchemexpress, Tyrphostin AG30 CAS:122520-79-0 Purity:>98%
SDP
Catalog No. 501969505
Encompass_Preferred
Medchemexpress, HY-118532 5mg Tyrphostin AG30 CAS:122520-79-0 Tyrphostin AG30 (AG30) is a potent and selective EGFR tyrosine kinase inhibitor. Tyrphostin AG30 (AG30) selectively inhibits self renewal induction by c-ErbB, and is able to inhibit activation of STAT5 by c-ErbB in primary erythroblasts. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9505 Supplier Medchemexpress LLC Supplier No. HY1185325MG
Medchemexpress, HY-118532 5mg Tyrphostin AG30 CAS:122520-79-0 Tyrphostin AG30 (AG30) is a potent and selective EGFR tyrosine kinase inhibitor. Tyrphostin AG30 (AG30) selectively inhibits self renewal induction by c-ErbB, and is able to inhibit activation of STAT5 by c-ErbB in primary erythroblasts. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.