Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tyrphostin AG490 | 134036-52-5 | MFCD00236452 | 99.7% | 294.30 | C17H14N2O3 | 1 ML
SDP

Catalog No. 5000240778
Encompass_Preferred

(E/Z)-AG490 is a racemic mixture of the E and Z isomers of AG490, a small-molecule tyrosine kinase inhibitor used in biochemical and cellular research. It is employed to probe JAK/STAT signaling and related pathways due to reported inhibition of JAK2/3, STAT3, and EGFR.

  • Racemic mixture of E and Z isomers.
  • Small-molecule tyrosine kinase inhibitor for signaling research.
  • Reported to inhibit JAK2/3, STAT3, and EGFR.
  • Available as a concentrated solution for cell-based assays (10 mM in DMSO).
  • High purity, 99.71%.
  • Molecular weight 294.30 and formula C17H14N2O3.
  • Offered in multiple solid and solution formats for assay flexibility.

Catalog No. 50-002-40778 Supplier Medchemexpress LLC Supplier No. HY10745910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E/Z)-AG490 is a racemic mixture of the E and Z isomers of AG490, a small-molecule tyrosine kinase inhibitor used in biochemical and cellular research. It is employed to probe JAK/STAT signaling and related pathways due to reported inhibition of JAK2/3, STAT3, and EGFR.

  • Racemic mixture of E and Z isomers.
  • Small-molecule tyrosine kinase inhibitor for signaling research.
  • Reported to inhibit JAK2/3, STAT3, and EGFR.
  • Available as a concentrated solution for cell-based assays (10 mM in DMSO).
  • High purity, 99.71%.
  • Molecular weight 294.30 and formula C17H14N2O3.
  • Offered in multiple solid and solution formats for assay flexibility.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.