Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (+)-Tyrphostin B44 (Tyrphostin AG 835) | 133550-37-5 | MFCD00209855 | 97.0% | 308.33 g/mol | C18H16N2O3 | 5 MG
SDP

Catalog No. 5000263699
Encompass_Preferred

(+)-Tyrphostin B44 (Tyrphostin AG 835) is a small-molecule research compound that acts as an epidermal growth factor receptor (EGFR) inhibitor and has reported antitumor activity. It is provided as a high-purity solid intended for in-vitro and preclinical research, with recommended storage conditions for powder and solutions to maintain stability.

  • EGFR inhibitor with reported antitumor activity.
  • High purity (97.04%) suitable for research applications.
  • Solid powder form for accurate weighing and formulation.
  • Recommended storage: powder at -20°C for long term or 4°C for short term.
  • Molecular weight 308.33 g/mol; formula C18H16N2O3.

Catalog No. 50-002-63699 Supplier Medchemexpress LLC Supplier No. HY101007A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(+)-Tyrphostin B44 (Tyrphostin AG 835) is a small-molecule research compound that acts as an epidermal growth factor receptor (EGFR) inhibitor and has reported antitumor activity. It is provided as a high-purity solid intended for in-vitro and preclinical research, with recommended storage conditions for powder and solutions to maintain stability.

  • EGFR inhibitor with reported antitumor activity.
  • High purity (97.04%) suitable for research applications.
  • Solid powder form for accurate weighing and formulation.
  • Recommended storage: powder at -20°C for long term or 4°C for short term.
  • Molecular weight 308.33 g/mol; formula C18H16N2O3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.