Learn More
Medchemexpress LLC Tyrphostin RG13022 | 136831-48-6 | 96.7% | 266.29 | C16H14N2O2 | 2 MG
SDP
Catalog No. 5000247653
Encompass_Preferred
RG13022 is a tyrosine kinase inhibitor that inhibits autophosphorylation of the epidermal growth factor receptor (EGFR) with an IC50 of 4 μM. It suppresses EGF-stimulated cancer cell proliferation and colony formation and has demonstrated tumor growth inhibition in vivo.
  • Inhibits EGFR autophosphorylation (IC50 4 μM).
  • Suppresses EGF-stimulated cell proliferation.
  • Demonstrated tumor growth inhibition in animal studies.
  • High purity suitable for research (≈96.7%).
  • Soluble in DMSO at 100 mg/mL for in vitro use.
Catalog No. 50-002-47653 Supplier Medchemexpress LLC Supplier No. HY1014292MG
RG13022 is a tyrosine kinase inhibitor that inhibits autophosphorylation of the epidermal growth factor receptor (EGFR) with an IC50 of 4 μM. It suppresses EGF-stimulated cancer cell proliferation and colony formation and has demonstrated tumor growth inhibition in vivo.
  • Inhibits EGFR autophosphorylation (IC50 4 μM).
  • Suppresses EGF-stimulated cell proliferation.
  • Demonstrated tumor growth inhibition in animal studies.
  • High purity suitable for research (≈96.7%).
  • Soluble in DMSO at 100 mg/mL for in vitro use.