Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tyrphostin RG13022 | 136831-48-6 | MFCD00236444 | >98.0% | 266.29 g/mol | C16H14N2O2 | 25 MG
SDP

Catalog No. 5000246788
Encompass_Preferred

RG13022 is a small-molecule tyrosine kinase inhibitor that blocks epidermal growth factor receptor (EGFR) autophosphorylation and suppresses EGF-stimulated cell proliferation in vitro; it also reduces tumor growth in vivo and is provided as a solid research reagent.

  • Inhibits EGFR autophosphorylation (IC50 ≈ 4 μM)
  • Molecular formula: C16H14N2O2
  • Molecular weight: 266.29 g/mol
  • Appearance: white to off-white solid
  • Typical application: biochemical and cell-based studies of EGFR signaling

Catalog No. 50-002-46788 Supplier Medchemexpress LLC Supplier No. HY10142925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RG13022 is a small-molecule tyrosine kinase inhibitor that blocks epidermal growth factor receptor (EGFR) autophosphorylation and suppresses EGF-stimulated cell proliferation in vitro; it also reduces tumor growth in vivo and is provided as a solid research reagent.

  • Inhibits EGFR autophosphorylation (IC50 ≈ 4 μM)
  • Molecular formula: C16H14N2O2
  • Molecular weight: 266.29 g/mol
  • Appearance: white to off-white solid
  • Typical application: biochemical and cell-based studies of EGFR signaling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.