Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ 400 Comfort Fit Design Coveralls, Open Wrists and Ankles
GREENER_CHOICE

Catalog No. 17500187 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-500-187 7X-Large
01-361-1B Medium
01-361-1C Large
01-361-1D X-Large
01-361-1E 2X-Large
01-361-1F 3X-Large
01-361-1G 4X-Large
01-361-1H 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 17-500-187 Supplier DuPont™ Supplier No. TY120SWH7X002500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Coveralls with collar and serged seams provide an ideal balance of protection, durability, and comfort of any limited use fabric technology.

  • Tyvek™ 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material.
  • Fabric offers an inherent barrier against particles (down to 1.0 micron in size).
  • Protection is built into the fabric itself, no films or laminates to abrade or wear away.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements.
  • Tailored fit, offers reinforcement in high stress areas for fewer blowouts.
  • Longer zipper for easier donning and doffing and an elastic waist to better position the garment.
  • Laydown collar, open wrists, open ankles.
  • Manufactured under specifications that do not contain natural rubber latex.
  • Stormflaps.

Compliance

Meets USDA, FDA, OSHA standards.

TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 7X-Large
Gender Unisex
Material Tyvek Flashspun HDPE
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Open
Cuff Style Open
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle
Collar Type Laydown
Certifications/Compliance ASTM D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127
Latex Free Yes
Model TY120SWH
For Use With (Application) Lead and Asbestos Abatement, Remediation, General Maintenance, Spray Painting, Cleaning
Quantity 25/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.