Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ 400 Coverall with Snap Closure
GREENER_CHOICE

Catalog No. 17500252 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-500-252 Large
17-500-249 2X-Large
17-500-250 3X-Large
17-500-251 4X-Large
17-500-253 Medium
17-500-254 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-500-252 Supplier DuPont™ Supplier No. TY151SWHLG002500
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Coveralls with collar provide an ideal balance of protection, durability, and comfort. Snap front design promotes easy donning and doffing.

  • Comfort fit design based on extensive wearer input to provide our most comfortable garment design that: enables a greater range of movement while stretching and bending, provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts, utilizes a longer zipper for easier donning and doffing and an elastic waist to better position the garm
  • Laydown collar
  • Snap front
  • Open wrists
  • Open ankles
  • Manufactured under specifications that do not contain natural rubber latex
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material Tyvek
Color White
Disposable Limited-use
Ankle Style Open
Cuff Style Open
Neck Style Collared
Hood Type Unhooded
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Collar Type Laydown
Certifications/Compliance ASTM D1777
Model TY151SWH
For Use With (Application) Lead and Asbestos Abatement, Remediation, General Maintenance, Spray Painting, Cleaning
Quantity 25/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.