Learn More
DuPont™ Tyvek™ 400 Coverall TY120S
Catalog No. 17111067 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
6X-Large
Catalog No. Size
17-111-067 6X-Large
1 options
Catalog No. 17-111-067 Supplier DuPont™ Supplier No. TY120SWH6X002500
Add to Cart
Add to Cart
Durable and lightweight coverall offering reliable protection, comfort fit design, and flexibility for industrial and maintenance use.
  • Garments are composed of flash spun high density polyethylene that forms a unique, nonwoven material available only from DuPont.
  • Provides an ideal balance of protection, durability, and comfort for limited use fabric technology.
  • Offers an inherent barrier against particles down to 1.0 micron in size.
  • Protection is built into the fabric itself with no films or laminates to abrade or wear away.
  • Delivers superior durability compared to microporous film fabrics, maintaining barrier integrity even after wear and abrasion.
  • Comfort fit design based on extensive wearer input allows greater range of movement while stretching and bending, offers a more tailored fit, provides reinforcement in high stress areas for fewer blowouts, includes a longer zipper for easier donning and doffing, and features an elastic waist for better garment positioning.
  • Includes a laydown collar, stormflap front, open wrists and ankles, and serged seams for enhanced strength and flexibility.
  • Pin locking slider locks securely in place when the pins on the puller are pushed into the zipper elements.
  • Manufactured under specifications that do not contain natural rubber latex.
  • Suitable for use in lead and asbestos abatement/remediation, general maintenance, spray painting, and clean-up applications.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Single-use Coveralls
Size 6X-Large
Gender Unisex
Material Spunbond Polypropylene
Color White
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Open
Cuff Style Open
Neck Style Collared
Hood Type Standard Collar
Closure Type Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Resistance Dust
Latex Free No
No. of Pockets None