Learn More
DuPont™ Tyvek™ 400 Lab Coat
GREENER_CHOICE
Catalog No. 17500265 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-500-265 2X-Large
17-500-267 3X-Large
17-500-269 4X-Large
17-500-270 5X-Large
17-500-274 Large
17-500-276 Medium
17-500-277 Small
17-500-279 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 17-500-265 Supplier DuPont™ Supplier No. TY212SWH2X0030NF
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Limited-use lab coats provide protection and durability. Meets USMCA standards.
  • Labcoat offers front snap closures for easy donning and doffing
  • Provides enhanced frontal protection
  • Laydown collar
  • Open wrists
  • Two front pockets for holding writing instruments, small tools or parts
  • Manufactured under specifications that do not contain natural rubber latex
Compliance
NAFTA
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Lab Coat
Size 2X-Large
Gender Unisex
Material Tyvek
Color White
Closure Type Snaps
Disposable Limited-use
Sterility Non-sterile
No. of Pockets 2
Model TY212SWH
Seam Type Serged