Learn More
DuPont™ Tyvek™ 400 Pants
GREENER_CHOICE
Catalog No. 17500290 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-500-290 2X-Large
17-500-291 3X-Large
17-500-292 4X-Large
17-500-296 Large
17-500-297 Medium
17-500-298 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-500-290 Supplier DuPont™ Supplier No. TY350SWH2X0012G1
Add to Cart
Add to Cart
This item is not returnable. View return policy
400 pants provide the protection and durability you need so you can focus on the task at hand. Meets USDA, FDA, OSHA Standards.
  • Pants with open ankle and elastic at waist for easy donning and removal
  • Suitable for applications that do not require the full body coverage of a coverall
  • Typically worn with TY303S WH shirt
  • Manufactured under specifications that do not contain natural rubber latex
  • Can be used in a variety of applications including general maintenance/operations, asbestos and lead abatement, mold remediation, and environmental cleanups
Compliance
USDA, FDA, OSHA
TRUSTED_SUSTAINABILITY
Specifications
Size 2X-Large
Gender Unisex
Material Tyvek
Color White
Ankle Style Open
Seam Type Serged
Waist Style Elastic
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.