Learn More
Description
- Pants with open ankle and elastic at waist for easy donning and removal
- Suitable for applications that do not require the full body coverage of a coverall
- Typically worn with TY303S WH shirt
- Manufactured under specifications that do not contain natural rubber latex
- Can be used in a variety of applications including general maintenance/operations, asbestos and lead abatement, mold remediation, and environmental cleanups
Compliance
USDA, FDA, OSHA
Specifications
Specifications
| Size | X-Large |
| Gender | Unisex |
| Material | Tyvek |
| Color | White |
| Ankle Style | Open |
| Seam Type | Serged |
| No. of Pockets | None |
| Waist Style | Elastic |
| For Use With (Application) | General Maintenance/Operations, Asbestos and Lead Abatement, Mold Remediation, Environmental Cleanups |
| Product Type | General Purpose Pants |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

