Learn More
DuPont™ Tyvek™ 400 Sleeve
GREENER_CHOICE
Catalog No. 17500301 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Universal
Catalog No. Size
17-500-301 Universal
1 options
Catalog No. 17-500-301 Supplier DuPont™ Supplier No. TY500SWH000200NF
  • $397.00
    $311.65 / Case of 200
    Save $85.35
    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Disposable sleeves provide the protection and durability to focus on the task at hand
  • Elastic at both ends (bicep and wrist) for arm protection
  • 18 in. long
  • Manufactured under specifications that do not contain natural rubber latex
  • One size fits most
  • D14352392
Compliance
NAFTA
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Sleeve
Size Universal
Material Tyvek
Color White
Sterility Non-sterile
Disposable Single-use
Closure Type Elastic Ends
Seam Type Serged
Length (English) 18 in.
Length (Metric) 45.72 cm
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.