Learn More
DuPont™ Tyvek™ 400 Pants
GREENER_CHOICE
Catalog No. 19820558 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-558 Medium
19-820-557 Large
19-820-559 X-Large
19-820-555 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-820-558 Supplier DuPont™ Supplier No. TY350SWHMD0050VP
Add to Cart
Add to Cart
This item is not returnable. View return policy
400 Pants come complete with an elastic waist and serged seams.
  • Pants with open ankle and elastic at waist for easy donning and removal
  • Suitable for applications that do not require the full body coverage of a coverall
  • Typically worn with TY303S WH shirt.
  • Manufactured under specifications that do not contain natural rubber latex
  • D15221896, D15221870, D15221862, D15221883
TRUSTED_SUSTAINABILITY
Specifications
Size Medium
Gender Unisex
Material Tyvek
Color White
Ankle Style Open
Seam Type Serged
Waist Style Elastic