Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ 400 SFR White Coverall ST120, Collared

Catalog No. 17900938 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-900-938 Medium
17-111-303 2X-Large
17-111-304 3X-Large
17-111-305 4X-Large
17-111-306 5X-Large
17-111-307 6X-Large
17-111-301 Large
17-111-302 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-900-938 Supplier DuPont™ Supplier No. ST120SWHMD002500
  • $414.00
    $262.65 / Case of 25

    Save $151.35

    37% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tyvek™ 400 SFR Coverall. Relaxed Fit Design. Collar. Storm flap. Red Zipper. Elastic Wrists and Ankles. Red External Serged Seams. White.

  • Garments are lightweight, disposable secondary flame-resistant overgarments designed to help protect and preserve primary flame-resistant garments, while providing an ideal balance of protection, durability, and comfort.
  • In the event that a flash fire occurs, Tyvek™ 400 SFR garments won't ignite and won't contribute to additional burn injury if appropriate primary flame-resistant personal protective equipment, such as DuPont™ Nomex™, is worn beneath.
  • Composed of flash spun high density polyethylene breathable nonwoven Tyvek™ fabric that has been flame-retardant treated, available only from DuPont, Tyvek™ 400 SFR garments offer an inherent barrier against particles (down to 1.0 micron in size).
  • Tyvek™ 400 SFR represents a new generation of disposable secondary flame-resistant garment technology. Compared to traditional disposable secondary flame-resistant garments that have been available for years, Tyvek™ 400 SFR is a differentiated multi-hazard garment solution, offering superior particle barrier protection and improved durability in a lighter-weight fabric.
  • Can be recycled through the Tyvek™ protective apparel recycling program if not contaminated with hazardous materials.
  • Applications include: oil and gas, general manufacturing/operations, maintenance, dirty jobs, environmental services, utilities.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Flame-resistant Coveralls
Size Medium
Gender Unisex
Material Tyvek
Color White
Disposable Reusable
Waist Style Standard
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Standard Collar
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Flame
No. of Pockets None
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.