Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ 400 SFR White Coverall ST120, Standard Fit Hood

Catalog No. 17200857 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-857 2X-Large
17-200-858 3X-Large
17-200-859 4X-Large
17-200-860 5X-Large
17-200-861 6X-Large
17-200-862 7X-Large
17-111-309 Large
17-111-308 Medium
17-111-310 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-200-857 Supplier DuPont™ Supplier No. ST127SWH2X002500
Add to Cart
Add to Cart

Tyvek™ 400 SFR Coverall. Relaxed Fit Design. Standard Fit Hood. Storm flap. Red Zipper. Elastic Wrists and Ankles. Red External Serged Seams. White.

  • Garments are lightweight, disposable secondary flame-resistant overgarments designed to help protect and preserve primary flame-resistant garments, while providing an ideal balance of protection, durability, and comfort.
  • In the event that a flash fire occurs, Tyvek™ 400 SFR garments won't ignite and won't contribute to additional burn injury if appropriate primary flame-resistant personal protective equipment, such as DuPont™ Nomex™, is worn beneath.
  • Composed of flash spun high density polyethylene breathable nonwoven Tyvek™ fabric that has been flame-retardant treated, available only from DuPont, Tyvek™ 400 SFR garments offer an inherent barrier against particles (down to 1.0 micron in size).
  • Tyvek™ 400 SFR represents a new generation of disposable secondary flame-resistant garment technology.
  • Can be recycled through the Tyvek™ protective apparel recycling program if not contaminated with hazardous materials.
  • Applications include: oil and gas, general manufacturing/operations, maintenance, dirty jobs, environmental services, utilities.
  • Relaxed fit coverall design features a diamond gusset crotch to allow for unrestricted movement and optimal fit when wearing primary flame-resistant protective clothing beneath
  • Red trim elements - red external stitched seams, red zipper, and red chest patch border
  • Patent-pending design
  • Attached standard-fit hood with elastic around face opening
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Storm flap
TRUSTED_SUSTAINABILITY

Specifications

Product Type Flame-resistant Coveralls
Size 2X-Large
Gender Unisex
Material Tyvek
Color White
Disposable Reusable
Waist Style Standard
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Flame
No. of Pockets None
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.