Learn More
DuPont™ Tyvek™ 500 Hooded Coverall
Catalog No. 17500258 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
4X-Large
5X-Large
6X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-500-258 Small
17-500-255 4X-Large
17-500-256 5X-Large
17-500-257 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 17-500-258 Supplier DuPont™ Supplier No. TY198SWHSM0025LA
Add to Cart
Add to Cart
This item is not returnable. View return policy
DuPont™ Tyvek™ 500 coveralls offer enhanced protection againt a range of threats, from waterbased chemical liquid splashes, asbestos, pharmaceutical and radioactive particulates to biological hazards.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Products made after January, 2023 are manufactured under specifications that do not contain natural rubber latex.
  • Protection against infective agents (EN 14126) including resistance to penetration by blood and body fluids using synthetic blood (ISO 16603 Class 3) and resistance to penetration by blood-borne pathogens using Phi-X174 bacteriophage (ISO 16604 Class 2)
  • Chemical protective clothing, Category III, Type 5-B and 6-B. (1956)
  • EN 1073-2 (protection against radioactive contamination)
  • Antistatic treatment (EN 1149-5) - on both sides.
  • Serged external seams featuring patent-pending technology for enhanced liquid spray tightness
  • Very low inward leakage due to optimized design
  • Tyvek™ auto-lock zipper and storm flap for increased protection
  • Elastic face, wrists and ankles as well as glued-in waist elastic
Certifications
Chemical Protective Clothing, Cat.III,Type 5B,6B
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protective Coveralls
Size Small
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Waist Style Elastic (Glued-in)
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Chemicals
No. of Pockets None
Style Storm Flap